VWR®, Measuring Beakers with Handle, PP
Supplier: VWR Collection
213-0497EA 1.7 GBP
213-0497 213-0498 213-0499 213-0500 213-0501 213-0502 213-0503
VWR®, Measuring Beakers with Handle, PP
Beakers
Made of PP, translucent.
  • With printed graduations
  • With handle for better usage
  • Withstand temperatures of 0 to 121 °C
  • Resin meets US FDA 21 CFR requirements
Order Now
SPECIFICATIONS
Environmentally Preferable Sustainable Packaging