Retort clamp
Supplier: R L ENTERPRISES
Z0104
212-9503EA 54.5 GBP
212-9503
Retort clamp
Clamps General Purpose Clamps
Alloy die-cast, aluminium shaft. This clamp accepts articles up to 85 mm diameter. Actuated by nickel-plated brass pummels and blue plastic headed nickel-plated steel twin screw.
  • Fitted with an aluminium shaft
  • Four interlocking jaws are cork lined