VWR®, Test Tube Racks, PP
Supplier: VWR Collection
212-0438EA 13.1 GBP
212-0438 212-0437 212-0436 212-0435 212-0434 212-0433 212-0432 212-0431
VWR®, Test Tube Racks, PP
Racks Tube Racks
PP.
  • Single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
Order Now
SPECIFICATIONS
Environmentally Preferable