Starch, soluble, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals

Synonyms: Starch, Vitex® iodine indicator, Amylodextrin

21153.135EA 20.1 GBP
21153.135 21153.181 21153.237 21153.290 21153.460
Starch, soluble, AnalaR NORMAPUR® zA
Starch, soluble
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878

Order Now

Specification Test Results

Appearance White fine powder
pH (2 %) 5,5 - 7,5
Sensitivity Passes test
Ignition residue Max. 0.4 %
Loss on drying (105°C; 2 h) Max. 10 %
Reducing substances (as C12H22O11) Max. 0.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR