Test tube and drying racks, Nalgene®
Supplier: Thermo Fisher Scientific
5977-0013 5977-0017 5977-0313 5977-0317
211-7211EA 31.8 GBP
211-7211 211-7221 211-7214 211-7224
Test tube and drying racks, Nalgene®
Racks Tube Racks
PP, reinforced.
  • Heat resistant up to +135 °C, autoclavable
  • Do not float in the water bath
  • Very good chemical resistance
Peg racks are ideal for holding sample collection tubes, reaction tubes and test tubes. The racks can also be used as drying racks for electrophoresis and chromatography plates and for a range of tubes.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603)