Strips of 8 PCR tubes with attached cap strips
Supplier: Brand
781330
211-3264EA 151 GBP
211-3264
Strips of 8 PCR tubes with attached cap strips
Tubes PCR Tubes
PP. Eight connected 0,2 ml tubes with attached domed cap strips.
  • Domed caps facilitate one-handed operation
  • Free from DNase, RNase and DNA
Colour: Clear
Colour: Clear