VWR®, Test Tube Racks, Fold and Snap
Supplier: VWR Collection
211-0351EA 13 GBP
211-0351 211-0352 211-0353 211-0354 211-0355 211-0356 211-0357 211-0358 211-0359 211-0360 211-0361 211-0362
VWR®, Test Tube Racks, Fold and Snap
Racks Tube Racks
PP.
  • Easy to assemble polypropylene racks snap together in seconds
  • Designed to remain stable in water baths
  • Shipped flat
Order Now
SPECIFICATIONS
Environmentally Preferable