VWR®, Racks, Microcentrifuge Tube
Supplier: VWR Collection
211-0213EA 55.6 GBP
211-0213 211-0214 211-0215 211-0216 211-0217 211-0218
VWR®, Racks, Microcentrifuge Tube
Racks Tube Racks
PP.
  • For freezer use
  • Side handles for secure handling
  • Numbered for easy sample identification
For 96 (8×12) microtubes with 0,5 ml capacity or 96 microtubes with 1,5/2 ml capacity. Conical, tapered openings to hold tubes securely. Complete with snap-on lid, which can also be used as a protective base.
Ordering information: The assorted colour kit includes one rack each coloured blue, green, pink, yellow and orange.