VWR®, Organisers for Microcentrifuge Tube Racks
Supplier: VWR Collection
211-0211EA 282 GBP
211-0211 211-0212
VWR®, Organisers for Microcentrifuge Tube Racks
Racks Tube Racks
Clear acrylic.
  • Stable, sturdy construction, space saving and stackable
  • Safe storage with quick and easy access to microcentrifuge racks
  • Easy to clean
Organiser for up to eight racks, designed to hold VWR® microcentrifuge tube racks. These racks help you keep large numbers of microtubes organised on your bench top. Up to eight 80-well microtube racks filled with either 1,5 or 2,0 ml microtubes.
Order Now
SPECIFICATIONS
Environmentally Preferable