VWR®, Culture and Sample Tubes, Disposable
Supplier: VWR Collection
211-0085EA 144 GBP
211-0085
VWR®, Culture and Sample Tubes, Disposable
Tubes Test Tubes
Transparent PS or translucent PP.
  • PS tubes: Not autoclavable. Temperature resistant from −40 to +90 °C. Can be centrifuged up to 1500×g
  • PP tubes: Autoclavable up to 121 °C. Temperature resistant from −90 to +135 °C. Can be centrifuged up to 3000×g
  • Press in stopper or dual position push on cap available for pressure equalisation or gas exchange whilst ensuring that the cap cannot fall off
  • Sustainable design based on virgin resins
Precision moulded culture and sample tubes for a wide variety of applications. With round bottom. Sterile or non sterile.