2,4,6-Trimethylphenylmagnesium bromide 1 M THF, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Mesitylmagnesium bromide, 0

210731000
210731000.EA 246 GBP
210731000.
2,4,6-Trimethylphenylmagnesium bromide 1 M THF, AcroSeal®
2,4,6-Trimethylphenylmagnesium bromide
Formula: C₉H₁₁BrMg
MW: 223.396 g/mol
Density: 1.005 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00009663
CAS Number: 2633-66-1
UN: 2924
ADR: 3 II

Order Now

Specification Test Results

Appearance Light yellow to brown Solution
Titration Grignard 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR