Benzyl alcohol ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

20805.291EA 82.9 GBP
20805.291
Benzyl alcohol ≥98%, GPR RECTAPUR®
Benzyl alcohol
Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205
Melting Pt: –15,3 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 101
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %
Boiling point 204 - 208 °C
Density (20/4) 1.043 - 1.046


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR