trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 99,0

Supplier: VWR Chemicals
Warning

Synonyms: 2,2',2'',2'''-(trans-Cyclohexane-1,2-diylbis(azanetriyl))tetraacetic acid hydrate, DCTA monohydrate, CDTA monohydrate, trans-Cyclohex-1,2-ylenediaminetetra acetic acid monohydrate, (1,2-Cyclohexylenedinitrilo)tetraacetic acid monohydrate, 2,2',2'',2'''-[trans-Cyclohexane-1,2-diylbis(azanetriyl)]tetraacetic acid hydrate, CHELATON 4, Titriplex® IV, trans-1,2-Cyclohexanediaminetetraacetic acid monohydrate, trans-Cyclohexane-1,2-dinitrilotetraacetic acid monohydrate

20284.153EA 132 GBP
20284.153
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 99,0
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Formula: C₁₄H₂₂N₂O₈·1H₂O
MW: 364.35 g/mol
Melting Pt: 213…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00066429
CAS Number: 125572-95-4
EINECS: 236-308-9

Order Now

Specification Test Results

Assay (calculated on anhydrous) Min. 99.0 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.1 %
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 10 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR