Fast Blue RR, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Warning

Synonyms: 4-Benzoylamino-2,5-dimethoxyaniline, Azoic Diazo No. 24, Azoic Diazo Component 24 (Base)

201545-5G
201545-5GEA 54.2 GBP
201545-5G
Fast Blue RR, Sigma-Aldrich®
Fast Blue RR

Fast Blue RR has been used to detect alkaline phosphatase activity.

Formula: C₁₅H₁₆N₂O₃
MW: 272.30 g/mol
Melting Pt: 165…168 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00008369
CAS Number: 6268-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1703210da3077c3e7d841b14168f930351bbb87&vid=09e02dfa62d083dc3410685741894ecf1832bb48&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR