Diisobutylaluminium hydride 1.0 M heptane, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

201051000 201058000
201051000.EA 145 GBP
201051000. 201058000.
Diisobutylaluminium hydride 1.0 M heptane, AcroSeal®
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Melting Pt: -80
Density: 0.798
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Clear solution
Strength 19.5 to 20.5 % (DIBAL-H, on Al basis)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR