Joint clips for spherical joints, POM, Quickfit®
Supplier: SciLabware
KS13 KS19 KS29 KS35
201-8179EA 46 GBP
201-8179 201-8180 201-8181 BIBBKS35
Joint clips for spherical joints, POM, Quickfit®
Clamps NS Joint Clamps
Clips made of polyacetal (POM) for retaining glassware having Quickfit® spherical joints.
  • Easy to fit and remove
  • Can be used at temperatures up to 80 °C
  • Clips are colour-coded for easy identification