Nitrogen flasks, round bottom (Schlenk Flasks) with NS stopcock
Supplier: Lenz Laborglas GmbH & CO.KG
03360122 03360128 03360137 03360437 03360449 03360458 03360470 03360428 03360149
201-1130EA 88 GBP
201-1130 201-1131 201-1132 201-1135 201-1136 201-1137 201-1138 LENZ03360428 LENZ03360149
Nitrogen flasks, round bottom (Schlenk Flasks) with NS stopcock
Flasks Nitrogen Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With conical ground joint, NS stopcock and inscription label.
Order Now
SPECIFICATIONS
Environmentally Preferable