Adapters, bends
Supplier: Lenz Laborglas GmbH & CO.KG
05025004 05025003 05025002 05025001
271-2203EA 58.6 GBP
271-2203 201-0571 201-0570 LENZ05025001
Adapters, bends
Distillation Components Distillation Adapters
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 90°. With 2 standard ground cones.