Acetic acid 96% pharmaceutical production

Supplier: VWR Chemicals
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

20093.320EA 76.9 GBP
20093.320 20093.460
Acetic acid 96% pharmaceutical production
Acetic acid

This product is prepared using glacial acetic acid Ph.Eur., USP and water purified Ph.Eur., USP, JP.


Caution: Not for use as Active Pharmaceutical Ingredient (API)

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3) II
REACH: 01-2119475328-30

Order Now

Specification Test Results

Assay ≥ 96.1 %
Appearance Clear colourless liquid
Identification Passes test
Reducing substances Passes test Ph.Eur.
Evaporation residue ≤ 0.02 %
Formic acid, formates (as HCOOH) (*) ≤ 0.5 %
Heavy metals (as Pb) ≤ 5 ppm
Cl (Chloride) ≤ 25 ppm
SO₄ (Sulphate) ≤ 50 ppm
Ca (Calcium) ≤ 5 ppm
Fe (Iron) ≤ 5 ppm
(*) Formic acid, formates, other oxidable impurities (as HCOOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR