Acetonitrile 99,5, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA

Supplier: VWR Chemicals
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

20071.294EA 60.3 GBP
20071.294 20071.328 20071.460 20071.550 20071.950
Acetonitrile 99,5, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45,7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.5 %
IR Spectrum Passes test
Solution (100 g/l) Passes test
Acidity Max. 0.0002 meq/g
Alkalinity Max. 0.0006 meq/g
Boiling point 80 - 82 °C
Colouration Max. 10 APHA
Density (20/4) 0.781 - 0.784
Density (20/20) 0.782 - 0.785
Refractive index (20°C) 1.343 - 1.345
Evaporation residue Max. 10 ppm
Water Max. 0.15 %
CN (Cyanide) Max. 50 ppm
Cd (Cadmium) Max. 0.02 ppm
Co (Cobalt) Max. 0.01 ppm
Cu (Copper) Max. 0.04 ppm
Fe (Iron) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Zn (Zinc) Max. 0.1 ppm
Transmittance (255-420 nm) Min. 98 %
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR