Smart Station ultrapure water remote dispenser, Arium®
Supplier: Sartorius
H2O-ARST-UP-T H2O-ARST-UP-B
171-1396EA 2370 GBP
171-1396 171-1395
Smart Station ultrapure water remote dispenser, Arium®
Water Dispensers
The Arium® Smart Station can be optimised to fit easily into any lab environment. Combine the Smart Station with the water system type needed to dispense pure or ultrapure water. The Smart Station ultrapure supports multiple points of use to save costs and valuable lab space.
  • Compact: Save space integrating into any lab
  • Intuitive: Touch-activated colour display with direct access to all important functions
  • Flexible: Stepless height adjustment to fill different size containers
  • Ergonomic design: Hand-held for optimised left- or right-hand operation
  • Accurate:
More Product Information