VWR®, Ultrapure Water System, A Series
Supplier: VWR Collection
171-1250EA 6020 GBP
171-1250 171-1249 171-1460 171-1461
VWR®, Ultrapure Water System, A Series
Water Purification Systems Pure and Ultra Pure Water Systems
This compact 'plug and play' lab water system is an ideal choice for users who need up to 20 litres of ultrapure Type I water or RO water (Type III) per day. Quality of the ultrapure product water meets or exceeds ASTM, CLSI, CAP, and ISO Type I water standards.
  • Simple and intuitive to dispense water via the dispenser or central console
  • Central console is an 8 inch waterproof touch screen, providing user full control and access of system performance
  • One water system can drive several dispensing units ('1 + N mode')
  • Dispensing flow rate is up to 0,5 L/min
More Product Information
Limited Time Sale
Select items below are currently on sale