Cartridges
Supplier: ELGA LabWater
LC111 LC127 LC128 LC148 LC129 LC182 LC183 LC149 LC185 LC150 LC184
ELGALC111EA 709 GBP
ELGALC111 171-0464 171-0465 171-0478 171-0466 171-0360 171-0361 ELGALC149 171-0363 ELGALC150 171-0362
Cartridges
Filters Filter Cartridges
The Ultra is available in five versions that specifically meet the different needs of the laboratories. Ultra Scientific delivers ultrapure water continuously. Its 2-cartridge pack ensures a constant water supply that meets the most demanding applications of scientists. Ultra Ionic meets the requirements of applications as critical as those of chromatography or spectrophotometry by eliminating residual ionic and organic fractions from pretreated water. Ultra Analytic by integrating a 185 nm UV lamp produces water with an exceptionally low TOC level (2 ppb). Display of TOC level in ppb. UltraMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable