(±)-α-Terpineol 96

Supplier: Thermo Fisher Scientific
Warning

Synonyms: α-Terpineol, Terpineol, 2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol

16285.22 16285.36 16285.14
16285.22EA 49.2 GBP
16285.22 16285.36 16285.14
(±)-α-Terpineol 96
(±)-α-Terpineol

Purity: 96% CAS No.: 98-55-5 Molecular Formula: C10H18O Formula Weight: 154.24 Flash Point: 90°(194°F) Physical Form: Liquid Density: 0.930 Refractive Index: 1.4830 Appearance: Colourless to pale yellow Melting Point: 31-32° Boiling Point: 210-219° Merck Reference: 14,9171 MDL No.: MFCD00001557


A naturally-occuring monoterpene alcohol

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 210…219 °C
Melting Pt: 31…32 °C
Density: 0.930
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001557
CAS Number: 98-55-5
EINECS: 202-680-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR