Multi-gas detectors, GasAlert MicroClip X3
Supplier: BW TECHNOLOGIES
MCX3-XWHM-Y-EU MCX3-XW0M-Y-EU MCX3-XW00-Y-EU MCX3-X000-Y-EU MCX3-XWH0-Y-EU MCX3-X000-Y-UK MCX3-XW00-Y-UK MCX3-XW0M-Y-UK MCX3-XWHM-Y-UK MCX3-XWH0-Y-UK
151-0238EA 663 GBP
151-0238 151-0236 151-0235 151-0234 151-0237 151-0295 151-0296 151-0297 151-0299 151-0298
Multi-gas detectors, GasAlert MicroClip X3
Gas Detection Systems
The GasAlert MicroClip X3 provides protection from atmospheric gas hazards, especially in cold weather. Reflex™ Technology ensures sensor responsiveness to enhance safety, uptime and worker confidence.
  • Rechargeable extended battery for harsh temperatures
  • Flashing green IntelliFlash™ light to check compliance at one glance
  • Simultaneous monitoring of up to four gases and combustibles in percentage of lower explosive limit (LEL)
  • Battery lifetime of 18 hours and 12 hours in cold temperatures, charge in six hours
  • One button operation reducing training time
More Product Information