Surface active cleaning agent, decon® 90
Supplier: DECON LAB
D901 D905
148-0324EA 27.2 GBP
148-0324 148-0325
Surface active cleaning agent, decon® 90
Cleaning Agents Detergents and Surface Cleansers
A surface active cleaning agent and/or radioactive decontaminant, for laboratory, medical and specialised industrial applications. A concentrate that is suitable for ‘manual’ cleaning, or for use in an ultrasonic cleaning bath.
  • Biodegradable, phosphate-free and enzyme-free
  • Bactericidal
  • Highly alkaline >pH 13
Note: Not suitable for domestic use nor suitable for use on non-ferrous metals, notably aluminium and zinc, or on polycarbonate.
Certifications: decon® 90 satisfies the criteria in BS EN 1040 1997 for bacterial activity.
Caution: NotMore Product Information