Palladium standard solution, 10000 mg/L Pd in 20% hydrochloric acid (from Pd), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

14432.AD 14432.AE 14432.AP
14432.ADEA 276 GBP
14432.AD 14432.AE 14432.AP
Palladium standard solution, 10000 mg/L Pd in 20% hydrochloric acid (from Pd), standard for ICP, Specpure®
Standards Cationic Standards Palladium Standards

Molecular Formula: Pd in 20% HCl
Physical Form: Liquid
Odour: Characteristic
UN No.: 1789
MDL No.: MFCD03613603


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR