Erbium standard solution, 10000 mg/L Er in 5% nitric acid (from Er₂O₃), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

14409.AD 14409.AE 14409.AP
14409.ADEA 90.9 GBP
14409.AD 14409.AE 14409.AP
Erbium standard solution, 10000 mg/L Er in 5% nitric acid (from Er₂O₃), standard for ICP, Specpure®
Standards Cationic Standards Erbium Standards

Molecular Formula: Er₂O₃ in 5% HNO₃ Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010987


Certifications: Traceable to NIST

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00010987

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR