SlimLine PLW 7111 Laboratory Glasswashers
Supplier: MIELE
11875050 11875090 11875120 11875190 11879730 11875200 11875410 11875470 11879740 11879750 11879760
141-1595EA 22725.36 GBP
141-1595 141-1596 141-1597 141-1598 141-1602 141-1599 141-1600 141-1601 141-1603 141-1604 141-1605
SlimLine PLW 7111 Laboratory Glasswashers
Glassware Washing Machines
The PLW 7111 SlimLine laboratory washers offer a comprehensive and systematic solution for analytically pure reprocessing of laboratory glassware.
  • Smart Load Plus - for flexible use of wash cabinet space
  • EasyLoad - systematic efficiency and convenience
  • Space-saving SlimLine design
  • Freely programmable controls
  • Flexible reprocessing of laboratory glassware
  • Status display via chamber lighting
  • Connectivity-ready
The PLW 7111 series of SlimLine laboratory glasswashers provide performance and user-friendliness. This entirely redesigned washing system boastsMore Product Information