Cerium standard solution, 1000 mg/L Ce in 5% nitric acid (from Ce), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

13815.ADEA 45.6 GBP
13815.AD 13815.AE
Cerium standard solution, 1000 mg/L Ce in 5% nitric acid (from Ce), standard for ICP, Specpure®
Standards Cationic Standards Cerium Standards

Molecular Formula: Ce in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010924


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR