Praseodymium standard solution, 1000 mg/L Pr in 5% nitric acid (from Pr₆O₁₁), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

13812.AEEA 79.6 GBP
13812.AE
Praseodymium standard solution, 1000 mg/L Pr in 5% nitric acid (from Pr₆O₁₁), standard for ICP, Specpure®
Standards Cationic Standards Praseodymium Standards

Molecular Formula: Pr6O11 in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011174

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR