Ductless fume hoods, Paramount®
Supplier: LABCONCO
6963320 6963521 6963623
135-2023EA 0 GBP
135-2023 LBCP6963521 LBCP6963623
Ductless fume hoods, Paramount®
Fume Hoods and Enclosures Ductless Fume Hoods
Self-contained ductless enclosures allow safe handling of organic, formaldehyde, acid gas, sulphur compounds, or ammonia chemical fumes and vapours, as well as particulates. Air inside the enclosures passes through carbon filters, and in some cases additional HEPA particulate filters, before being safely exhausted back into the laboratory. No outside ducting is required.
  • Telescoping base stands adjust in seven height positions from 69,9 to 75,1 cm
  • Glacier white stands are constructed of 4,4 cm tubular epoxy-coated steel
  • Hydraulic lift base stands are
More Product Information