Rhodium 99,95 Metals basis, slug

Supplier: Thermo Fisher Scientific

13381.KGEA 0 GBP
13381.KG
Rhodium 99,95 Metals basis, slug
Rhodium
Formula: Rh
MW: 102.9 g/mol
Boiling Pt: 3400…3700 °C (1013 hPa)
Melting Pt: 1960
Density: 12.41 g/cm³ (20 °C)
MDL Number: MFCD00011201
CAS Number: 7440-16-6
EINECS: 231-125-0
Merck Index: 12,08353

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR