Calcium chloride dihydrate 99,0-105,0, granular, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride

1332-05EA 313 GBP
1332-05 1332-01 1332-07
Calcium chloride dihydrate 99,0-105,0, granular, BAKER ANALYZED® ACS, J.T.Baker®
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Boiling Pt: 1600
Melting Pt: 176
MDL Number: MFCD00149613
CAS Number: 10035-04-8
Merck Index: 13,01660

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CaCl₂) 74.0 - 78.0 %
Assay (CaCl₂·2H₂O) 99.0 - 105.0 %
Insoluble Matter ≤0.01 %
pH of 5% Solution at 25°C 4.5 - 8.5
Oxidizing Substances (as NO₃) ≤0.003 %
Sulfate (SO₄) ≤0.01 %
Ammonium (NH₄) ≤0.005 %
Barium (Ba) ≤0.005 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.02 %
Strontium (Sr) ≤0.1 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR