Blue Super-Duty Autoclavable SMS Muff Covers
Supplier: KEYSTONE CLEANROOM PRODUCTS
MFKCRP600-05 MFKCRP600-07 MFKCRP600-03 MFKCRP600-06 MFKCRP600-09 MFKCRP600-18 MFKCRP600-12 MFKCRP600-04
129-4020EA 1730 GBP
129-4020 129-4022 KACCMFKCRP600-03 KACCMFKCRP600-06 KACCMFKCRP600-09 KACCMFKCRP600-18 KACCMFKCRP600-12 KACCMFKCRP600-04
Blue Super-Duty Autoclavable SMS Muff Covers
Covers
This 88 g/m² SMS autoclavable muff cover provides an excellent microbial barrier that is ideal for pharmaceutical production applications.
  • Eliminates autoclave tape residue
  • Hydrophobic, tear-resistant, and puncture-resistant
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Designed for autoclave cycles (121 to 135 °C)
  • Manufactured and packed in a certified ISO 8 cleanroom
  • Used in conjunction with a secondary autoclave bag, it creates an effective wrapping system
  • Made in USA
Eliminate the need for cutting sheetsMore Product Information