Platinum, black, HiSPEC® 1000

Supplier: Thermo Fisher Scientific

Synonyms: Platinium

12755.03 12755.06 12755.14 12755.01
12755.03EA 0 GBP
12755.03 12755.06 12755.14 12755.01
Platinum, black, HiSPEC® 1000
Platinum

CAS No.: 7440-06-4 Physical Form: S.A. nominally 27m2/g Odour: Odourless MDL No.: MFCD00011179

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1 II
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR