Alcalase (from Bacillus licheniformis)
Supplier: Merck Millipore Calbiochem
126741-500
126741-500EA 234 GBP
126741-500
Alcalase (from Bacillus licheniformis)
Enzymes
A serine endopeptidase that consists primarily of subtilisin A. Suitable for the hydrolysis of proteins.
One unit is defined as the amount of enzyme that will release 1,0 µmol L-tyrosine from hemoglobin per min at 25 °C.