IQ/OQ/PQ Kit for Ion Chromatography, certified reference material, Supelco®
Supplier: Merck
12674-1KT
12674-1KTEA 309 GBP
12674-1KT
IQ/OQ/PQ Kit for Ion Chromatography, certified reference material, Supelco®
Standards Anionic Standards Multi-Element Anioinic Standards
IQ/OQ/PQ-Kit for the qualification of ion chromatography systems. Suitable for the verification of linearity, repeatability and carry-over.
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034
Packaging: Contains six certified bromide standards (5, 10, 20, 50, 100 and 1000 mg/kg; 100 ml each). The exact contents, traceability and uncertainty are given in a detailed certificate.