Potassium carbonate 99,0 ACS

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Potash

12609.22 12609.36 12609.A1 12609.A7 12609.14
12609.22EA 34.6 GBP
12609.22 12609.36 12609.A1 12609.A7 12609.14
Potassium carbonate 99,0 ACS
Potassium carbonate

Purity: 99.0% min CAS No.: 584-08-7 Molecular Formula: K2CO3 Formula Weight: 138.21 Physical Form: Granular Density: 2.29 Melting Point: 891° Beilstein Reference: 4267587 Merck Reference: 14,7619 MDL No.: MFCD00011382

Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891
Density: 2.29
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 14,7619
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR