Calcium chloride dihydrate 99

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride

12312.36 12312.A3
12312.36EA 61.2 GBP
12312.36 12312.A3
Calcium chloride dihydrate 99
Calcium chloride dihydrate

Purity: 99% min CAS No.: 10035-04-8 Molecular Formula: CaCl2.2H2O Formula Weight: 147.02 (110.99anhy) Physical Form: Granular Density: 0.835 Merck Reference: 14,1659 MDL No.: MFCD00149613

Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Boiling Pt: 1600
Melting Pt: 176
Density: 0.835
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR