(±)-1,2-Propanediol, Supelco®

Supplier: Merck

Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane

MQ100

12279-1ML-F 12279-5ML-F
12279-1ML-FEA 46.9 GBP
12279-1ML-F 12279-5ML-F
(±)-1,2-Propanediol, Supelco®
Propylene glycol
Formula: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Boiling Pt: 187.6
Melting Pt: –59 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 99
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
EINECS: 200-338-0
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64030690e2aa187bd981b8997eec130abe165797&vid=479429a8062338eaedf8207d99f0c46cd7d8d474&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR