Iron(III) nitrate nonahydrate 98 Metals basis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate

12226.22 12226.A1 12226.A7
12226.22EA 23.5 GBP
12226.22 12226.A1 12226.A7
Iron(III) nitrate nonahydrate 98 Metals basis
Iron (III) nitrate nonahydrate

Purity: 98+% CAS No.: 7782-61-8 Molecular Formula: FeN3O9.9H2O Formula Weight: 403.99 (241.86anhy) Physical Form: Crystalline, Contains excess HNO3 Density: 1.684 Appearance: Pale purple Odour: Acidic Melting Point: 47° Boiling Point: 125° dec. Merck Reference: 14,4027 UN No.: 1466 MDL No.: MFCD00149708

Formula: Fe(NO₃)₃·9H₂O
MW: 404 g/mol
Boiling Pt: 125 °C (dec.)
Melting Pt: 47
Density: 1.684
Storage Temperature: Ambient
MDL Number: MFCD00149708
CAS Number: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1 III
Merck Index: 14,4027

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR