Nickel(II) nitrate hexahydrate Ni ≥ 19,8%, tech.

Supplier: Thermo Fisher Scientific
Danger

12222.36 12222.A3
12222.36EA 43.1 GBP
12222.36 12222.A3
Nickel(II) nitrate hexahydrate Ni ≥ 19,8%, tech.
Nickel (II) nitrate hexahydrate

CAS No.: 13478-00-7 Molecular Formula: N2NiO6.6H2O Formula Weight: 290.80 (182.70anhy) Physical Form: Flake Density: 2.05 Sensitivity: Hygroscopic Appearance: Green Odour: Odourless Melting Point: 56.7° Boiling Point: 137° Solubility: Soluble in water, alcohol Merck Reference: 14,6513 UN No.: 2725 MDL No.: MFCD00149805


Nickel plating, manufacturing of ceramic Colours

Formula: Ni(NO₃)₂·6H₂O
MW: 290.79 g/mol
Boiling Pt: 137 °C
Melting Pt: 56.7
Density: 2.05
Storage Temperature: Ambient
MDL Number: MFCD00149805
CAS Number: 13478-00-7
EINECS: 236-068-5
UN: 2725
ADR: 5.1 III
Merck Index: 14,6513

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR