Cesium carbonate ≥99.994% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Warning

12117.22 12117.06 12117.14
12117.22EA 306 GBP
12117.22 12117.06 12117.14
Cesium carbonate ≥99.994% (metals basis), Puratronic®
Cesium carbonate

Purity: 99.994% CAS No.: 534-17-8 Molecular Formula: Cs2CO3 Formula Weight: 325.82 Physical Form: Powder/Granules Density: 4.072 pH: 10-12 Melting Point: 610° dec. Beilstein Reference: 4546405 Merck Reference: 14,2010 MDL No.: MFCD00010957

Formula: Cs₂CO₃
MW: 325.82 g/mol
Melting Pt: 610 °C (dec.)
Density: 4.072
Storage Temperature: Ambient
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9
Merck Index: 14,2010

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR