Spill containment units
Supplier: NEW PIG
PAK735-BK-WOD
121-0184EA 1800 GBP
121-0184
Spill containment units
Spill Control Materials
Spill containment units made from LDPE.
  • Catch spills if containers leak, drip or crack
  • Low profile for easy access to and dispensing from tanks
  • Resists UV rays, rust, corrosion and most chemicals
If this product is used with the flammable liquids, please consider the regulations that apply to storage and handling of flammable liquids and the safety of this application, specifically flammable vapours, static discharge and heat sources.