Bio-bin® Waste Disposal Boxes
Supplier: ECONIX LTD
BEXBB5RD BEXBBLOOP BEXBB5TS BEXBB5YL BEXBB5OR BEXBB2TS BEXBB2YL BEXBB1RD BEXBB1OR BEXBB1YL BEXBBLOOPOR BEXBB1MW BEXBB1TS BEXBB30YL BEXBBLOOPYL BEXBB1PL BEXBB30TS BEXBB30RD BEXBB30PL BEXBB30OR BEXBB30MW BEXBB30 BEXBB5PL BEXBB2RD BEXBB5MW BEXBB2OR MPEXBB1TS MPEXBB1OR MPEXBB1.YL MPEXBB1RD MPEXBB2TS MPEXBB2OR MPEXBB2.YL MPEXBB2RD MPEXBB5TS MPEXBB5OR MPEXBB5.YL MPEXBB5MW MPEXBB5RD MPEXBB5PL BEXBB30PHARMA
119-0224EA 424 GBP
119-0224 119-0482 119-0225 119-0176 119-0208 119-0215 119-0174 119-0214 119-0205 119-0173 119-0483 119-0213 119-0212 119-0178 119-0175 119-0211 119-0221 119-0220 119-0219 119-0207 119-0218 119-0470 119-0222 119-0217 119-0223 119-0206 119-0473 119-0471 119-0467 119-0472 119-0476 119-0474 119-0466 119-0475 119-0481 119-0478 119-0465 119-0477 119-0480 119-0479 ECONBEXBB30PHARMA
Bio-bin® Waste Disposal Boxes
Boxes Waste Disposal Boxes
These U.N. approved, paper-based containers provide a safe and innovative way to dispose all kinds of waste.
  • Space efficient fold-flat design with easy assembly
  • Carbon footprint reduction
  • Ideal for use in labs, wards and for community health
The 1 litre Bio-bin® is ideally suited for use in the field (e.g. community nursing) or for private first aid providers. It is the ideal size to be used in laboratories where limited space is available, such as inside fume cupboards.
The 2 litre Bio-bin® has been designed to replace plastic waste containers forMore Product Information