Nickel(II) perchlorate hexahydrate, Reagent Grade

Supplier: Thermo Fisher Scientific
Danger

11631.14 11631.22 11631.36
11631.14EA 28.8 GBP
11631.14 11631.22 11631.36
Nickel(II) perchlorate hexahydrate, Reagent Grade
Nickel (II) perchlorate hexahydrate

CAS No.: 13520-61-1 Molecular Formula: Ni(ClO4)2.6H2O Formula Weight: 365.70 (257.59anhy) Physical Form: Damp crystalline solid Refractive Index: 1.55 Sensitivity: Hygroscopic Appearance: Green Odour: Odourless Melting Point: 140° Solubility: Very soluble in water. Soluble in many organic solvents UN No.: 1481 MDL No.: MFCD00149804

Formula: Cl₂NiO₈·6H₂O
MW: 365.68 g/mol
Melting Pt: 140
Storage Temperature: Ambient
MDL Number: MFCD00149804
CAS Number: 13520-61-1
EINECS: 237-124-1
UN: 1481
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR