Industrial label printer, BradyPrinter i3300
Supplier: Brady
241132 241133 149556 149555 241120 241121 241126 241127
116-0819EA 1206.27 GBP
116-0819 116-0820 116-0808 116-0807 116-0811 116-0812 116-0815 116-0816
Industrial label printer, BradyPrinter i3300
Printers Label Printers
Print any label or sign with the versatile BradyPrinter i3300 industrial label printer.
  • Wire and cable identification labels
  • Product and laboratory identification labels
  • Safety and facility signs
The BradyPrinter i3300 Industrial Label Printer is a completely hassle-free, easy to use PC-based printer. It can print a full range of safety and facility labels and signs as well as die-cut labels for wire, product and laboratory identification.
With the BradyPrinter i3300, you can print a huge range of reliable labels and signs at your location inMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Display Display colour LCD touchscreen, 272×480 px
Label Continuous tape and die cut labels
Memory 512 MB (RAM), 195 MB (data storage), internal
Resolution 300 dpi
Dimensions 240W×317H×435D mm
Weight 5.30 kg