Silicone bench protector
Supplier: DELTALAB
350002
115-9137EA 142 GBP
115-9137
Silicone bench protector
Surface Protectors Bench Protectors
Silicone sheet designed for laboratory benchtop protection or for use in autoclaves to protect any material.
  • Easy to clean with a wet cloth
  • Heat resistant up to 260 ºC