Cleanroom wipes, ThermaSeal™ 60
Supplier: Texwipe
TX2069 TX2068B TX2069B TX2064 TX2064B TX4392
115-0466EA 94.5 GBP
115-0466 115-1888 115-1889 115-1887 TEXWTX2064B TEXWTX4392
Cleanroom wipes, ThermaSeal™ 60
Wipes Cleanroom Wipes Dry Cleanroom Wipes
ThermaSeal™ wipes are hot cut polyester wipes for all-purpose cleaning in critical environments. Ideal for cleaning medical devices, displays and products in the semiconductor and microelectronics area.
  • Thermally sealed edges to minimise fibre generation
  • Excellent absorbency, good abrasion resistance
  • Chemically compatible with standard solvents and alcohols
  • Autoclavable
Colour: White
Data accurate at time of publishing, latest test results and test methods can be found on supplier’s website.
Packaging: Solvent-safe Bag-Within-A-Bag® cleanroomMore Product Information